Hydrochloric acid 0.5 N volumetric solution, traceable to NIST Standard Reference Material (SRM)

Supplier: VWR
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

ANAC46498-700
71007-506CS 375.09 CAD
71007-506 71007-508
Hydrochloric acid 0.5 N volumetric solution, traceable to NIST Standard Reference Material (SRM)
Hydrochloric acid
Formula: HCl
MW: 36.46
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Normality 0.495 - .0505 N
Traceable to NIST standards


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR