Anti-IgY Donkey Polyclonal Antibody (CF™ 488A)

Supplier: Biotium

20166 20166-1
71003-936EA 522.35 CAD
71003-936 71003-938
Anti-IgY Donkey Polyclonal Antibody (CF™ 488A)
Antibodies
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: CF™ 488A
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Chicken
Order Now

SPECIFICATIONS

Antigen symbol IgY
Antigen name Immunoglobulin Y
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation CF® 488A
Reactivity Chicken
Host Donkey
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Epitope Heavy and Light chain
Cross adsorption Minimal Cross-Reactivity: Bv, Gt, GP, Hs, Hu, Ms, Rb, Rt, Sh, SHm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR