Low TOC vials, C&G Containers
Supplier: C&G Containers
LFV040700005 LFV040700500
71001-334CS 322.31 CAD
71001-334 71001-336 76423-308
Low TOC vials, C&G Containers
Vials
These glass vial are used primarily for the collection and testing of aqueous samples for Total Organic Carbon (TOC). Secondary uses include containment of extracts. Please refer to the applicable certificate of analysis for certified analytes and corresponding reporting limits.
  • Certificate of analysis and cleaning protocol form included in each product case
  • Cast PTFE thin septa cap with dust covers
For pre-cleaned and certified products, a lot certificate of analysis is generated with the analytical results obtained including the concentrationMore Product Information