Plastic Packers, Wide Mouth, HDPE, Natural, C&G Containers
Supplier: C&G Containers
LPN128900002 LPW002200010 LPW002200011 LPW002900000 LPW004200011 LPW004900000 LPW008200012 LPW008220012 LPW008230012 LPW008900000 LPW008900002 LPW016200010 LPW016200012 LPW016220012 LPW016230012 LPW016900002 LPW032200013 LPW032230013 LPW032900003 LPW064200010 LPW064900004 LPW066200010 LPW128200010 LPW128900000
71001-010CS 46.33 CAD
71001-010 71001-106 71001-108 71001-110 71001-114 71001-116 71001-118 71001-122 71001-124 71001-128 71001-130 71001-132 71001-134 71001-136 71001-138 71001-140 71001-142 71001-144 71001-146 71001-148 71001-152 71001-158 71001-160 71001-162 71001-164 71001-166 71001-170 71001-174
Plastic Packers, Wide Mouth, HDPE, Natural, C&G Containers
Bottles Water Sampling Bottles
These plastic bottles are used for the collection of aqueous media for testing metals and a variety of inorganic parameters, including alkalinity, anions, acidity, cyanide, sulfate, and hardness.
Pre-cleaned and Certified products contain the Certificate of Analysis and Cleaning Protocol Form in the product case.
A lot certificate of analysis is generated with the analytical results obtained, including the concentration reporting limits and units of measurement, for Pre-cleaned and Certified products. This certificate of analysis is included in each product case.
Wash protocols forMore Product Information