I-CHEM™ Clean Snap Vials with 0.125" Septa, Thermo Scientific
Supplier: Thermo Scientific
SS136-0040 SS246-0040 SS236-0040
71000-080CS 437.75 CAD
71000-080 71000-084 71000-086 71000-088 71000-090 71000-082
I-CHEM™ Clean Snap Vials with 0.125" Septa, Thermo Scientific
Vials
Protect samples from contamination during storage or transport with Thermo Scientific™ Clean snap clear vials.
An innovative snap-off cap reduces the chance of volatile organic contaminants in the environment from passing through the septum while also keeping the septum clean and free from foreign particles during handling. The clean snap cap easily peals or snaps away to provide access to the septum at the time of analysis.
Certifications: Certified to meet or exceed EPA performance-based standards for volatile organic analysis.
Order Now
SPECIFICATIONS
Environmentally Preferable