SP Bel-Art Fritware® Porous Rods, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F13635-0012 F13635-0014
70690-266EA 204.71 CAD
70690-266 70690-268
SP Bel-Art Fritware® Porous Rods, Bel-Art Products, a part of SP
Retort Rods
Porous 30.5 cm (12") polyethylene rods can withstand temperatures up to 93°C (200°F).
  • Polyethylene construction
  • Porous design
  • Coarse porosity around 110 microns
Coarse grade porosity with an average pore size of 110 microns.