SP Bel-Art Cover Glass Forceps, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H37942-0000 H37943-0000
70680-086PK 133.01 CAD
70680-086 70690-198
SP Bel-Art Cover Glass Forceps, Bel-Art Products, a part of SP
Forceps
These forceps are fully coated with brown Teflon® containing FEP and comply with FDA regulations concerning direct food contact.
  • FEP coating
  • Wide tips
  • Bent or straight tips design
Wide tips are perfect for lifting cover glasses. Available in two styles, bent or straight tip.
Order Now
SPECIFICATIONS
Environmentally Preferable