SP Bel-Art Microscope Slide Tray and Cabinet, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H44583-0000 H44581-0000
70680-536EA 803.28 CAD
70680-536 70690-018
SP Bel-Art Microscope Slide Tray and Cabinet, Bel-Art Products, a part of SP
Slide Holders
Cabinet holds 12 Microscope Trays horizontally in a dust-free enclosure.
  • Polypropylene outer shell construction
  • Clear acrylic door with knob
  • Handy cut-out simplifies slide removal
The cabinet features a white polypropylene outer shell, polystyrene tray rails, and a vertically sliding, clear acrylic door with a knob for easy opening.
Each tray stores 20 standard 25 x 75 mm (1 x 3") slides in a horizontal position for full visibility. The white polystyrene background makes it easy to distinguish dyes and stains. Trays can be used independently, orMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable