SP Bel-Art Biohazard Incinerator Cartons, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F13205-0002 F13205-0001
70680-350PK 172.85 CAD
70680-350 70680-352
SP Bel-Art Biohazard Incinerator Cartons, Bel-Art Products, a part of SP
Boxes Waste Disposal Boxes
This corrugated cardboard receptacle is perfect for containing biohazardous waste slated for incineration.
  • Corrugated cardboard construction
  • Contains biohazardous waste without exposing users
  • Bright, large biohazard graphics printed on side
  • Optional reusable cover
When the box is full, pull the flap to a closed position and the entire unit can be placed in the incinerator. Tabs along the top edge keep a clear 0.038 mm (1.5 mil) thick polyethylene biohazard bag from slipping out of place and the snug-fitting lid has a flap for easily inserting wasteMore Product Information