Sodium dodecyl sulfate (SDS) ≥85% Ph. Eur., BP, JP, NF

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

SO180-500GM SO180-45KGBL SO180-2.5KG SO180-12KG SO180-125GM SO180-1KG
95035-854EA 419.93 CAD
95035-854 89079-012 95035-852 700006-728 95035-848 95035-850
Sodium dodecyl sulfate (SDS) ≥85% Ph. Eur., BP, JP, NF
Sodium dodecyl sulphate (SDS)

Sodium Lauryl Sulfate, NF, EP, BP, JP is used as an emulsifying agent, modified-release agent, penetration enhancer, solubilizing agent, tablet and capsule lubricant. All Spectrum Chemical NF, EP, BP, and JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: 204…207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 2811
ADR: 6.1 III
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 85% Min.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR