Potassium thiocyanate ≥98.5%, crystals ACS

Supplier: Spectrum Chemical Mfg. Corp.
Warning

P1420-500GM
700002-034EA 981.12 CAD
700002-034
Potassium thiocyanate ≥98.5%, crystals ACS
Potassium thiocyanate

Potassium Thiocyanate, Crystal, Reagent, ACS, one of the pseudohalides, is an important salt of the thiocyanate anion. Often used in chemical synthesis, it can also be used in theatrical special effects to create a relatively realistic blood effect. As an ACS grade Reagent, Spectrum Chemical manufactured potassium thiocyanate is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness

Formula: KSCN
Boiling Pt: 500 °C (1013 hPa)
Melting Pt: 175 °C
Density: 1.61 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1
Merck Index: 12,07866

Order Now

Specification Test Results

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR