20 mm Headspace Vials, Thermo Scientific
Supplier: Thermo Scientific
C4020-410 C4020-S10 C4020-S20
66065-340CS 2009.55 CAD
66065-340 66065-344 66065-348 66065-358 69400-116 69400-118 66030-782 66030-784 66064-346 66064-350 66065-328
20 mm Headspace Vials, Thermo Scientific
Vials
Thermo Scientific National Headspace Vials meet or exceed OEM specifications.
Vials feature a beveled crimp finish which gives a much tighter seal than flat crimp surfaces. Manufactured from clear 33 expansion borosilicate glass. Flat-bottom vials feature a slightly longer neck, providing a custom fit for Carlo Erba and Agilent® autosamplers. Round-bottom vials are designed to fit PerkinElmer®, Tekmar®, and Varian® headspace autosamplers. Seals have attached septa, unless otherwise specified.
Thermo Scientific National Unassembled Headspace Convenience Kits are available for the mostMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable