PYREX® Watch Glasses, Corning
Supplier: Corning
9985-65 9985-75 9985-90 9985-100 9985-125 9985-150
66110-021CS 891.83 CAD
66110-021 66110-043 66110-065 66110-087 66110-101 66110-123
PYREX® Watch Glasses, Corning
Watch Glasses
PYREX® borosilicate glass.
  • High chemical durability
  • Prevents contamination of solutions
These watch glasses are molded to the same radius of curvature for convenient stacking. The heavy wall and uniform fire-polished edge provides the mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected.
They are also used as covers for heated beakers.
Packaging: One package has 12 watch glasses.
Order Now
SPECIFICATIONS
Environmentally Preferable