Staining Jar with Lid, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
276840
66012-440CS 764.52 CAD
66012-440
Staining Jar with Lid, WHEATON®, DWK Life Sciences
Jars Staining Jars
The WHEATON Coplin Jar has an extra tall lid to accept 5 3" x 1" (75mm x 25mm) or 3" x 1-1/2 (100mm x 25mm) slides vertically or up to 10 back-to-back
The jar and cover is manufactured from soda-lime glass, and the jar has an approximate inside bottom dimensions of 28mm L x 26mm W x 92 mm H.