Sample Vials, Screw Thread, Borosilicate Glass, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
60810-1940 60910-1 60910-2 60910-3 60910-4 60910-6 60910-8 60910C-12 60910C-2 60910C-4 60910C-6 60910C-8 60957-1 60957-4 60910D-1 60910D-2 60910D-3 60910D-4 60957C-4 60957C-6 60957C-11 60810-1528 60910C-1 60910C-112 60910C-3 60910L-12 60910L-1 60910L-2 60920D-4 60920D-8 60957D-4 60957D-6 60957D-11 60815-1760 60815-1965 60920D-1 60810-1760 60810-1965 60810-2170 60810-2385 60811S-5 60811S-10 60812B-1 60812B-12 60812B-1232 60812B-2 60812B-3 60812B-4 60812B-6 60912-1545 60912-1235 60912-1760 60912-1528 60912-1738 60912-1940 60912-2857 60912-2895 60912D-1 60912D-12 60912D-2
66009-813CS 1193.44 CAD
66009-813 66010-243 66010-247 66010-249 66010-251 66010-253 66010-255 66010-260 66010-268 66010-272 66010-274 66010-276 66010-431 66010-437 66010-442 66010-445 66010-449 66010-451 66012-140 66012-142 66012-146 66009-816 66010-265 66010-267 66010-270 66010-286 66010-300 66010-322 66010-408 66010-410 66010-446 66010-448 66010-452 66010-482 66010-484 66010-522 66010-560 66010-564 66010-570 66010-581 89201-774 89201-776 89201-804 89201-806 89201-808 89201-810 89201-812 89201-814 89201-816 89201-850 89201-852 89201-854 89201-856 89201-858 89201-860 89201-862 89201-864 89201-874 89201-876 89201-878
Sample Vials, Screw Thread, Borosilicate Glass, Kimble Chase, DWK Life Sciences
Vials
General-purpose, clear and amber borosilicate glass vials.
Volumes range from 0.5 to 11 drams.
Certifications: Meets USP Type I and ASTM E438, Type I, Class A and B requirements.
Ordering information: Available with or without screw caps. Caps are packed separately.