Tygon® A-60-G Chemical Dispensing Tubing
Supplier: Saint Gobain Life Sciences
AFL00003 AFL00008 AFL00012 AFL00018 AFL00019 AFL00020 AFL00022 AFL00027 AFL00029 AFL00032 AFL00038
62993-014PK 121.6 CAD
62993-014 62993-018 62993-019 62993-023 62993-024 62993-026 62993-056 62993-057 62993-058 62993-059 62993-061 62993-089 62993-092 62993-093 62993-114 62993-115 62993-118 62993-148 62993-163 62993-172
Tygon® A-60-G Chemical Dispensing Tubing
Tubing Transfer Tubing
Tygon® A-60-G is a thermoplastic elastomer black tubing which will not weaken or crack after years of aging, heat, or ozone.
Chemical compatibility approaches that of fluoroelastomer and silicone tubings. Resistant to ammonia, bleach, brake fluid, bromine, chlorine, essential oils, ethylene glycol, formaldehyde, hydrochloric acid, hydrogen peroxide, iodine, methanol, nitric acid (10%) plating solutions, soap solutions, sodium hydroxide (to medium concentrations), and sulfuric acid (to medium concentrations).
Offers superior resistance to flex-fatigue in peristaltic pumps and is heat-More Product Information