Corning® 3-Hole Delivery Cap, High Temperature, Corning
Supplier: Corning
1395-45DC
62890-448CS 568.18 CAD
62890-448
Corning® 3-Hole Delivery Cap, High Temperature, Corning
Closures Bottle Closures
Red polybutylene terephthalate (PBT) GL45 screw cap has three 6.4mm (1/4") holes at top for use with 3.2mm (1/8") O.D
tubing. Caps have fluoropolymer resin liner for efficient sealing and may be heat sterilized at up to 180°C (356°F).
Order Now
SPECIFICATIONS
Environmentally Preferable