Nalgene® Mini Hose Barb Connectors; PP, Thermo Scientific
Supplier: Thermo Fisher Scientific
7210-0500
62860-554CS 353.43 CAD
62860-554
Nalgene® Mini Hose Barb Connectors; PP, Thermo Scientific
Fittings and Connectors
Nalgene Mini Hose Barb Polypropylene Connectors allow connections between 3/4" sanitary fitting to flexible tubing
CAUTION! Plastic tanks are generally subject to more severe conditions than plastic labware; exposure is constant, stresses are greater and different classes and concentrations of chemicals are involved. Please pay special attention to chemical compatibility.
Comes with an inner diameter of 1/2".