Louvered Wall Panel, Akro-Mils
Supplier: Akro-Mils
30636
62661-490CS 1049.89 CAD
62661-490
Louvered Wall Panel, Akro-Mils
Furniture Backsplashes, Wallracks, Wallpanels
Wall-mounted louvered panels keep parts up off the ground and free up floor space.
  • Formed from 16-gauge cold-rolled steel for strength and durability
  • Durable powder-coated finish stands up to heavy-duty service
  • Single-sided
For use with AkroBins or InSight Bins (sold separately).