Corning® Cell Culture Roller Bottles, Corning
Supplier: Corning
431133 431135 431329 431198 431191 3907 431321 431134 430195 430699 430849 430851 430852 430853 431344
21008-660CS 422.44 CAD
21008-660 21008-662 22250-132 25382-462 29442-914 33500-694 33502-074 46610-074 62404-370 62404-440 62404-442 62404-444 62404-446 62404-448 89089-838
Corning® Cell Culture Roller Bottles, Corning
Bottles Roller Bottles
Virgin PS, sterile.
  • One piece seamless construction
  • All bottles have printed lot numbers to aid product traceability
  • Sterilized by gamma radiation
  • Certified non-pyrogenic
  • Ribbed design provides twice the surface area with the same exterior dimensions
Roller bottles available with smooth or ribbed surface design for expanded culture area. All bottles are treated for optimal cell attachment. The Corning® CellBIND® surface enhances cell attachment under difficult conditions, such as reduced-serum or serum-free medium, resulting in higher cell yields. AllMore Product Information