Microtiter® 96-Well Fluorescence Microplates, Thermo Scientific
Supplier: Thermo Fisher Scientific
7905 7805 7705 7605
62402-980CS 661.9 CAD
62402-980 62402-983 62403-150 62403-156
Microtiter® 96-Well Fluorescence Microplates, Thermo Scientific
Microplates HTS Microplates
These 96-well polystyrene microplates are designed to optimize assay performance in fluorescent applications when used with light emitting markers
Black microplates reduce background signal to improve assay sensitivity, reduce crosstalk, and avoid light piping between wells. White microplates offer higher reflectivity to enhance signal and reduce crosstalk for increased dynamic range. High-binding microplates are irradiated. Plates with U-shaped bottoms have a 0.28mL well capacity; flat-bottom wells accommodate 0.33mL.
Plates are compatible with Thermo Scientific universal microplateMore Product Information