Celstir® Suspension Culture Flasks with Jacketed Double Sidearm, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
356946 356949 356952 356954 356943 356945
62401-964CS 1513.38 CAD
62401-964 62401-966 62401-968 62401-970 62401-960 62401-962
Celstir® Suspension Culture Flasks with Jacketed Double Sidearm, Wheaton
Flasks Spinner Flasks
Designed for suspension cell culture with a magnetic stirrer.
  • Inlet and outlet hose barb connections for 0.25” (6.35 mm) ID tubing
  • 360° water jacket for precise temperature control
  • Autoclavable borosilicate glass
Certifications: Conforms to USP Type I and ASTM E 438 Type 1, Class A requirements
Order Now
SPECIFICATIONS
Environmentally Preferable