Celstir® Suspension Culture Flasks, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
356876 356879 356882 356887 356889 356873 356875 356884 356890
62401-904CS 784.82 CAD
62401-904 62401-906 62401-908 62401-912 62401-914 62401-900 62401-902 62401-910 62401-916
Celstir® Suspension Culture Flasks, Wheaton
Flasks Spinner Flasks
For quality laboratory production and trypsinizing suspended cell cultures
Flask design provides an optimum inert cell growth environment. An adjustable blade impeller maximizes incubator space and provides maximum turbulence. A bottom dimple (on 125mL and larger sizes) prevents crushing of cells beneath the magnetic impeller and improves cell yield. Units larger than 250mL have 45mm sidearms, which can be used as air vents, media inlet/outlet ports, inoculation port, or to accommodate a pH probe. The screw cap assembly provides a positive seal using a fluoropolymer resin liner backedMore Product Information