Container for Used Blades, Martor
Supplier: Martor
9810.08
61200-328EA 41.13 CAD
61200-328
Container for Used Blades, Martor
Bins Sharps Containers
You can fill this container with the content of your Safebox. Or directly with your used blades.
When the box is full, turn the lid until it shuts. Now the box cannot be reopened. The next step is to safely and completely dispose of the filled Used Blade Container as normal waste.