Nalgene® Racks, Polycarbonate, Thermo Scientific
Supplier: Thermo Fisher Scientific
5929-0016 5929-0020 5929-0030
60985-137CS 744.7 CAD
60985-137 60985-148 60985-159
Nalgene® Racks, Polycarbonate, Thermo Scientific
Racks Tube Racks
Racks are ideal for use below 4°C (39.2°F)
Note: Autoclavable, but repeated autoclaving may reduce mechanical strength.
They are made of transparent, rustproof, and corrosion-resistant polycarbonate. Holes are keyed with numbers and letters for easy identification. Wide spacing allows convenient insertion and removal of tubes. Racks stack when empty for convenient storage. Will not float in water baths.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO14001 (Certificate Registration No. EMS 752603)