Anti-Signal peptide peptidase-like 2B (SPPL2b) Rabbit Polyclonal Antibody

Supplier: Abgent
AP6311A
60942-670EA 1205.56 CAD
60942-670
Anti-Signal peptide peptidase-like 2B (SPPL2b) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Signal peptide peptidase-like 2B (SPPL2b)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR