Anti-CDKN2A (Cyclin-dependent kinase inhibitor 2A) Rabbit Polyclonal Antibody

Supplier: Abgent
AP3186A
60942-584EA 1199.09 CAD
60942-584
Anti-CDKN2A (Cyclin-dependent kinase inhibitor 2A) Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDKN2A (Cyclin-dependent kinase inhibitor 2A)
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR