Anti-CREBBP Rabbit Polyclonal Antibody

Supplier: Abgent
AP1077B
60941-186EA 1205.56 CAD
60941-186
Anti-CREBBP Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-CREBBP (Center) Reacts with Human

Type: Primary
Antigen: CREBBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR