SP Bel-Art No-Wire™ Test Tube Racks, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
187450000 187450002 F18745-0003 187460000 F18746-0001 F18746-0002 187460003 187470000 187470001 187470003 F18747-0004 F18745-0004 187460004 10153-666
60915-400CS 397.37 CAD
60915-400 60915-404 60915-405 60915-406 60915-408 60915-410 60915-411 60915-412 60915-414 60915-417 60915-418 60915-422 60915-425 10153-666
SP Bel-Art No-Wire™ Test Tube Racks, Polypropylene, Bel-Art Products, a part of SP
Racks Tube Racks
A great alternative to wire racks, with price- and space-saving advantages over other plastic racks.
  • Blend of polypropylene construction
  • Autoclavable
  • Meets the demands of daily usage in the lab
  • Available in numerous hole sizes
  • Choice of three colors for safety and identification
  • Features end plates for labeling; numbered and lettered rows for tube identification
  • Interlocking feet allow stacking for storage
  • Steam autoclavable at 121 °C (250 °F)
Made of a special blend of autoclavable polypropylene; will sink in a water bath and maintain stability, evenMore Product Information