LabSolutions™ Detergents, Labconco®
Supplier: Labconco
4422000 4422100 4522000 4522200
21851-130EA 292.3 CAD
21851-130 21851-132 60787-680 60787-682
LabSolutions™ Detergents, Labconco®
Cleaning Agents Detergents and Surface Cleansers
Formulated for use in automatic laboratory glassware washers
Low-Foaming Liquid Detergent removes a wide range of laboratory contaminants. Noncorrosive formula is safe for use on stainless steel, glass, or plastic. Phosphate-free for environmentally safe disposal and chlorine-free for safe use tissue cultures. Can be used in hot or cold water. Comes in a 3.8L (1gal.) jug. Washes approximately 180 loads.
Non-Foaming Powder Detergent effectively removes grease, blood, agar, and protein digestates from glassware. Special formula prevents damage to machine pumps, seals, and gaskets. A 1%More Product Information