Nalgene® Heavy-Duty Cylindrical Tanks with Spigot, LLDPE, Thermo Scientific
Supplier: Thermo Fisher Scientific
11102-0005 11102-0007 11102-0010 11102-0015 11102-0030 11102-0055
60464-021EA 592.23 CAD
60464-021 60464-027 60464-032 60464-043 60464-065 60464-087
Nalgene® Heavy-Duty Cylindrical Tanks with Spigot, LLDPE, Thermo Scientific
Tanks Cylindrical Tanks
Heavy-duty construction provides extra rigidity so the tanks hold shape well over time.
Equipped with polypropylene needle spigot, stepped flange at rim, and overlapping skirted polyethylene cover. Graduated in gallons and liters, with the exception of tanks 60464-027 and 60464-032, which are graduated in gallons only. Good chemical resistance and tolerant of a wide range of temperatures. Not autoclavable.
Order Now
SPECIFICATIONS
Environmentally Preferable