Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific
Supplier: Thermo Fisher Scientific
14200-0010 14200-0002 14200-0005 14200-0020 14200-0015
60463-104EA 717.88 CAD
60463-104 60463-105 60463-107 60463-113 73470-028
Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific
Tanks Rectangular Tanks
Tanks have stepped rim for strength and overlapping skirted cover
Polypropylene tank is molded in one piece and is autoclavable
Certifications: Plastic meets USP VI and FDA 21 CFR 177.1520 standards.
Ordering information: Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.
Order Now
SPECIFICATIONS
Environmentally Preferable