GC Septa, Hamilton Company
Supplier: Hamilton
76022 76010 76003 76004 76005 76013 76015 76020 76016 75801 75810 75804 75803 75805 75806 75807 75813 76001 76002 76006 76009 76017 76019 76007
60379-501PK 28.29 CAD
60379-501 60379-545 60379-589 60379-603 60379-625 60379-669 60379-749 60379-862 60379-760 60379-884 60379-909 60379-964 89220-870 89220-872 89220-874 89220-876 89220-878 89220-880 89220-882 89220-884 89220-886 89220-890 89220-892 89220-894 89220-888
GC Septa, Hamilton Company
Septa
Hamilton Company offers the most comprehensive selection of products in the industry.
  • Offers a variety of septa for many GC
  • Each is manufactured to achieve the highest level of quality possible
High temperature septa is made of preconditioned PTFE/silicone rubber and is 3.175 mm (0.125") thick. The average life of these septa is up to 100 injections with a 26s gauge needle. The maximum operating temperature is 212 °C.
Three Layer Septa is made of preconditioned silicone rubber and is 3.81 mm (0.150") thick. The average life of these septa is up to 1More Product Information