Gastight® Microlab® 500 and 1000 Instrument Syringes, Hamilton Company
Supplier: Hamilton
80222 80926 81026 81226 81326 80262 80962 81060 81062 81160 81260 81262 81660 80922 81022 81162 81222 201200 201250 201300 201350 201150 201050 201500 201600 81560 81460 80226 81126 81122 81323 201100 81360 201000 201400 80062 CAL86020
25229-008EA 230.25 CAD
25229-008 25229-045 25229-078 25229-136 25229-170 25229-206 25229-210 25229-212 25229-214 25229-216 25229-218 25229-220 25229-222 25229-351 25229-384 25229-412 25229-442 60376-000 60376-004 60376-006 60376-008 60376-018 72310-018 72310-022 72310-024 72310-396 72310-398 25229-012 25229-103 25229-410 60376-290 72310-020 72310-400 89221-378 89221-384 89221-466 76437-220
Gastight® Microlab® 500 and 1000 Instrument Syringes, Hamilton Company
Syringes Syringes for Chromatography
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry. Each is manufactured to achieve the highest level of accuracy and precision possible.
Gastight® syringes are ideal for dispensing both liquids and gases
These syringes are designed to fit into or with various different instruments such as the Microlab® 500 and Microlab® 1000.
Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision-machined PTFE plunger tip which creates a leak-free seal. With the tight fit, the tip essentially wipes the interior ofMore Product Information