Gastight® 1700 Series Syringes, Hamilton
Supplier: Hamilton
80927 81027 81127 81227 80065 80265 80965 81065 81265 80920 81020 81120 81220 80901 81001 81101 81201 80030 80039 80230 80930 81030 81230 80000 80200 80900 81000 81100 81217 80239 80939 81139 81243 80075 80085 80275 80285 80985 81075 81175 81185 81216 80201 80001 81165 81130 81039 80975 81085 80016 80004 80050 80950 81050 80008 80208 80908 81008 81214
60373-852EA 176.58 CAD
60373-852 60373-854 60373-856 60373-858 60373-985 60373-987 60373-988 60373-990 60373-994 60375-373 60375-409 60375-431 60375-464 60376-026 60376-048 60376-060 60376-081 60376-172 60376-176 60376-183 60376-194 60376-208 60376-219 60376-220 60376-230 60376-241 60376-252 60376-263 60376-274 60376-510 60376-520 60376-540 60376-551 60376-652 60376-654 60376-656 60376-658 60376-662 60376-664 60376-668 60376-670 60376-672 72310-388 72310-390 60373-992 60375-442 60376-528 60376-660 60376-666 89210-100 89221-252 30617-744 30617-746 30617-748 10192-074 10192-076 10192-078 10192-080 10192-082
Gastight® 1700 Series Syringes, Hamilton
Syringes Syringes for Chromatography
These syringes are the Gastight® version of the original 700 series syringes and are ideal for dispensing volumes from 1μL up to 500μL.
Gastight® syringes are ideal for dispensing both liquids and gases and have a precision-machined PTFE plunger tip which creates a leak-free seal
Cemented needles for low volume syringes are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-More Product Information