Microliter™ Built-In Syringe Guide Syringes, Hamilton Company
Supplier: Hamilton
80307 88407 88007 80107 80337 80407 86207 88507 86254
60373-800EA 180.93 CAD
60373-800 60375-074 60375-095 60375-138 72310-438 89221-262 89221-296 89221-304 89221-298
Microliter™ Built-In Syringe Guide Syringes, Hamilton Company
Syringes Syringes for Chromatography
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
Each is manufactured to achieve the highest level of accuracy and precision possible. The syringes in this series have a built-in syringe guide installed by the factory that prevent the plunger from bending and pulling out of the syringe. Factory installation ensures the guide is applied correctly and eliminates inadvertent damage caused by incorrect do-it-yourself assembly. These 700 and 7000 series Microliter™ syringes have a stainless steel plunger that is hand fitted to the glassMore Product Information