GASTIGHT® 1000 Series Syringes, Hamilton
Supplier: Hamilton
81327 81427 81527 81627 82527 85027 82520 85020 86020 81301 81401 81501 81601 81317 81417 81517 81617 81430 81530 81630 81543 81343 81443 81616 81316 81416 81516 81322 81643 81365 81514 81330 203260 203270 85021 81314 81414 7649-01 7650-01 7651-01
60373-860EA 239.14 CAD
60373-860 60373-862 60373-864 60373-866 60373-868 60373-870 60375-624 60375-668 60375-670 60376-106 60376-128 60376-140 60376-161 60376-285 60376-300 60376-321 60376-354 60376-412 60376-423 60376-434 60376-438 60376-561 60376-565 60376-650 60376-674 60376-678 60376-680 72310-008 72310-392 82020-762 82020-764 60376-558 89210-072 89210-074 89221-472 10192-084 10192-086 10192-112 10192-114 10192-116
GASTIGHT® 1000 Series Syringes, Hamilton
Syringes Syringes for Chromatography
The 1000 series is a mid volume Gastight® syringe. Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision-machined PTFE plunger tip which creates a leak-free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze.
This series of syringes is ideal for dispensing volumes from 100μL up to 100mL
A variety of different terminations and needle options are available.More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable