SGE Inlert Liners, GC, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
092005 092008 092011 092013 092028 092031 092061 092064 092089 092098 092101 092111 092115 092117 092142 092148 092223 092272 092273 092048 092001 092002 092004 092007 092010 092016 092018 092019 092021 092022 092024 092027 092030 092034 092037 092062 092082 0920831 092085 0920861 092087 092091 092092 092097 092100 092103 092141 092144 092147 092150 092210 092211 092219 092220 092222 092224 092228 092229 092234 092235 092236 092244 092036 092045 092046 092065 092066 092095 092243 092068 092245 092038 092039 092084 092218 092017 092230 092003 09211301 09222125 09224525 092049 092058 092059 092071 092077 092155 0920990 09202501 09202601 09203825 09204625 09209225 09209401 09209901 09210025 09202101 09209525
14226-444PK 380.1 CAD
14226-444 14226-446 14226-448 14226-450 14226-454 14226-456 14226-458 14226-460 14226-464 14226-466 14226-468 14226-470 14226-474 14226-476 14226-478 14226-480 14226-482 14226-486 14226-488 14233-670 14233-678 60362-504 60362-506 60362-508 60362-510 60362-512 60362-514 60362-518 60362-520 60362-522 60362-524 60362-526 60362-528 60362-530 60362-532 60362-536 60362-548 60362-550 60362-554 60362-558 60362-562 60362-564 60362-566 60362-568 60362-572 60362-574 60362-576 60362-578 60362-580 60362-582 60362-584 60362-586 60362-588 60362-590 60362-592 60362-594 60362-596 60362-600 60362-602 60362-610 60362-612 60362-614 60362-618 60362-642 60362-644 60362-646 60362-648 60362-650 60362-652 60362-666 60362-672 60362-674 60362-690 60362-692 60362-702 87003-016 60362-516 60362-604 60362-664 10799-748 10799-750 10799-752 10799-828 10799-830 10799-832 10799-834 10799-836 10799-838 10799-844 10799-846 10799-848 10799-850 10799-852 10799-854 10799-856 10799-858 10799-860 10001-536 10001-538
SGE Inlert Liners, GC, Trajan Scientific and Medical
Chromatography Supplies Chromatography Inlet Liners
This wide range of fully deactivated inlet liners is compatibile with the industry's most popular instrument makes and models.
Liners cover a variety of applications, and ensure complete sample transfer from syringe to column without loss. The improved designs result in increased precision and accuracy of sample transfer.
Varian® 1177 injectors accept all Agilent Technologies 6.3 mm O.D. liners.
Order Now
SPECIFICATIONS
Environmentally Preferable