Thermo GC TriPlus Autosampler Syringes, SGE
Supplier: Trajan Scientific and Medical
000490 000492 002993 000790 0029841 0029931
60361-376EA 364.05 CAD
60361-376 60361-378 60362-192 89209-074 10799-780 10799-782
Thermo GC TriPlus Autosampler Syringes, SGE
Syringes Syringes for Chromatography
SGE Autosampler Syringes feature Diamond Syringe Technology
Enhancements to SGE Diamond Syringes result in improved solvent resistance, great temperature ranges and increased operational smoothness. These improvements deliver longer syringe life and an improved cycle life of up to ten times that of a similar competitor’s syringe. SGE Diamond Autosampler Syringes offer a new level of accuracy due to reduced carryover and no risk of adhesive contamination. Diamond Autosampler Syringes are color coded by volume for easy identification of installed syringes and are packaged in 100% recyclableMore Product Information