VWR® Talon® Cylinder Bench Clamps
Supplier: VWR International
97035-528 97035-530 60142-003 60142-006
97035-528EA 237.7 CAD
97035-528 97035-530 60142-003 60142-006
VWR® Talon® Cylinder Bench Clamps
Clamps General Purpose Clamps
Rugged cast aluminum cylinder clamps secure gas cylinders to benches, tables or other flat surfaces up to 6.4 cm (21/2") thick.
  • Rugged design
  • Cast aluminum construction
  • Non-slip spring catch and buckle
  • Easily adjustable
  • Dimensions: 8.3L x 13.3W x 16.5H cm (31/4 x 51/4 x 61/2")
The 2.5 cm (1") wide, 137.2 cm (54") long nylon strap features a nickel-plated, non-slip spring catch and buckle for easy adjustment around cylinders 10.2 – 35.6 cm (4 – 14") in diameter.
Order Now
SPECIFICATIONS
Environmentally Preferable