Synthware Vacuum Trap Assembly with #15 O-Ring Joint, Kemtech America
Supplier: Kemtech America
V391557
60004-656EA 740.52 CAD
60004-656
Synthware Vacuum Trap Assembly with #15 O-Ring Joint, Kemtech America
Vacuum Traps
Designed for use with manifolds that have a #15 O-ring joint.
  • Heavy wall Borosilicate construction
  • 35/20 Spherical joint
The seal between upper and lower parts is made with a 57 mm flat O-ring flange. The trap body has an O.D. of 57 mm, and is 300 mm in length. Complete trap assembly is supplied with horseshoe clamp, one #331 Viton® O-ring, and two #116 Viton® O-rings. All accessory items are sold separately.
Order Now
SPECIFICATIONS
Environmentally Preferable