Synthware Vacuum Manifold with High Vacuum Valves, Kemtech America
Supplier: Kemtech America
M202403 M202404 M202405 M522403 M522404 M522405
60004-538EA 980.06 CAD
60004-538 60004-540 60004-542 60004-544 60004-546 60004-548
Synthware Vacuum Manifold with High Vacuum Valves, Kemtech America
Manifolds
Vacuum manifolds are available with a serrated hose connection (10 mm ext. Ø) on the right, left, or both right and left hand sides.
  • Hi vacuum valves
  • Heavy wall borosilicate construction
  • 3, 4 or 5 port options available
The main tube has a 22 mm ext. Ø, and the distance between valves is 100 mm center to center. All manifolds feature a stopcock bore of 0 to 4 mm.