Synthware Condenser, Air Reflux, Threaded, Microscale, Kemtech America
Supplier: Kemtech America
C221410
60004-116EA 58.93 CAD
60004-116
Synthware Condenser, Air Reflux, Threaded, Microscale, Kemtech America
Condensers
Air reflux condenser for microscale applications.
  • Heavy wall borosilicate construction
  • Threaded upper joint
  • Open cap included
Supplied with a nylon compression cap and Viton® O-ring.