Synthware High Vacuum Valve, Teflon® Protected, 3-Way Metering
Supplier: Kemtech America
S640008
60004-040EA 237.66 CAD
60004-040
Synthware High Vacuum Valve, Teflon® Protected, 3-Way Metering
Valves Vacuum Valves
Viton® O-rings on PTFE shaft are protected with precision Teflon® wipers. The plug has a machined-in integral PTFE tip O-ring to make the primary seal.
  • Teflon® shaft
  • PTFE covered Viton® O-rings
Valves are designed to provide an excellent vacuum tight seal without exposing any material other than Teflon® to chemicals.