Synthware High Vacuum Valves, Teflon® Protected, Low Hold-Up
Supplier: Kemtech America
S630004 S630008 S630012
60004-032EA 133.67 CAD
60004-032 60004-034 60004-036
Synthware High Vacuum Valves, Teflon® Protected, Low Hold-Up
Valves Vacuum Valves
Valves feature two 9 mm O.D. sidearms bent to minimize the hold-up of liquids.
  • Heavy wall construction
  • 33 expansion Borosilicate glass
Viton® O-Rings on PTFE shaft are protected with precision Teflon® wipers. The plug has a machined-in integral PTFE tip O-Ring to make the primary seal. This design provides an excellent vacuum tight seal without exposing any material other than Teflon® to chemicals.