Synthware Powder Funnels, Kemtech America
Supplier: Kemtech America
F182450 F182475 F184100 F188100
60003-388EA 80.99 CAD
60003-388 60003-392 60003-396 60003-398
Synthware Powder Funnels, Kemtech America
Funnels Powder Funnels
Funnels feature a 60° side angle and an inner [ST] joint.
  • Ground glass joints
  • Heavy wall borosilicate construction
  • Various joint sizes available
These powder funnels are made of glass.