Synthware Flow Control Adapter, Chromatography, Rodaviss® Joint, Kemtech America
Supplier: Kemtech America
C432440
60001-748EA 176.24 CAD
60001-748
Synthware Flow Control Adapter, Chromatography, Rodaviss® Joint, Kemtech America
Adapters Adapters, Joints with Tube Connectors
This flow control adapter is designed for use with chromatography columns with a 24/40 or 45/50 inner Rodaviss® joint.
  • Open top screw cap, loosening cap, and Viton® O-ring included
  • With metering plug
Supplied with an open top screw cap, loosening cap, and Viton® O-ring. Adapter has a 0 to 4 mm metering plug.
Order Now
SPECIFICATIONS
Environmentally Preferable