Synthware Bushing Adapters, Kemtech America
Supplier: Kemtech America
A791419 A793519 A793524 A791924 A792434 A792445
60000-010EA 50.12 CAD
60000-010 60000-012 60000-014 60000-016 60000-020 60000-022
Synthware Bushing Adapters, Kemtech America
Adapters Adapters, Conical Ground Joints
Bushing adapters allow for compact assemblies of dissimilar joints.
  • Top outer [ST] joint
  • Bottom inner [ST] joint
The [ST] 10/18 or [ST] 10/30 sizes on small inner joints are useful as fittings for thermometers.