VWR® Solid Rubber Stoppers, Black
Supplier: VWR International
59580-047 59580-069 59580-080 59580-105 59580-127 59580-149 59580-160 59580-182 59580-207 59580-229 59580-240 59580-262 59580-284 59580-309 59580-342 59580-364 59580-386 59580-400 59580-422 59580-444 59580-466 59580-488 59580-502 59580-524 59580-546 59580-322
59580-047CS 1090.33 CAD
59580-047 59580-069 59580-080 59580-105 59580-127 59580-149 59580-160 59580-182 59580-207 59580-229 59580-240 59580-262 59580-284 59580-309 59580-342 59580-364 59580-386 59580-400 59580-422 59580-444 59580-466 59580-488 59580-502 59580-524 59580-546 59580-322
VWR® Solid Rubber Stoppers, Black
Stoppers Rubber Stoppers
These solid, black stoppers contain a high percentage of pure rubber, weigh less than neoprene, and are low in free sulfur (1/2%).
  • Resilient and pliable for a long time
  • Low in free sulfur
  • Can withstand adverse laboratory conditions without hardening
Composition ensures resiliency and pliability over a long period.
Packaging: Packaging Information: 53 stoppers per pack.
Order Now
SPECIFICATIONS
Environmentally Preferable