Deck-Mounted Needle Valve, WaterSaver Faucet
Supplier: WaterSaver Faucet
CT2880234WSA CT2880231WSA CT2880232AWSA CT2880232SWSA
VFCT2880234WSAEA 777.11 CAD
VFCT2880234WSA 59382-082 59382-084 59382-086
Deck-Mounted Needle Valve, WaterSaver Faucet
Valves Air Regulation Valves
White, epoxy-coated, single straight pattern laboratory needle valve.
  • Black, nylon, hooded handle with color-coded index button
  • Rated for use at pressures up to 150 psi
Brass construction with stainless steel needle and seat. Needle and seat are removable/replaceable for infinite service life. Mounting shank included.
Ordering information: Please specify service when ordering.
Order Now
SPECIFICATIONS
Environmentally Preferable